The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175911.3 | NP_787107.1 | 2 - 3 | 482 | 105 |
XM_011544169.2 | XP_011542471.1 | 2 - 3 | 226 | 105 | |
GenBank mRNA | BC028158.1 | - | 2 - 3 | 320 | 105 |