Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIER2
Entrez Gene ID
54531
Gene Name
MIER family member 2
Gene Aliases
KIAA1193, Mi-er2
Chromosome Location
Chr.19: 305573 - 344793 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017550.1 NP_060020.1 3 - 4 252 62
XM_005259582.4 XP_005259639.1 3 - 4 836 62
XM_006722769.3 XP_006722832.1 3 - 4 840 62
XM_011528072.2 XP_011526374.1 3 - 4 838 62
XM_011528073.1 XP_011526375.1 3 - 4 254 62
XM_011528074.2 XP_011526376.1 3 - 4 842 62
XM_011528075.2 XP_011526377.1 3 - 4 1359 62
XM_011528077.2 XP_011526379.1 4 - 5 3200 62
GenBank mRNA AB033019.1 - - 240 62
BC028203.1 - 3 - 4 252 62

View Full Product Details