The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015056.2 | NP_055871.1 | 7 - 8 | 731 | 103 |
GenBank mRNA | AK056263.1 | - | 7 - 8 | 694 | 103 |
BC014005.2 | - | 7 - 8 | 703 | 103 | |
BC028386.2 | - | 7 - 8 | 736 | 103 | |
D80001.1 | - | 7 - 8 | 629 | 103 |