Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIWIL1
Entrez Gene ID
9271
Gene Name
piwi like RNA-mediated gene silencing 1
Gene Aliases
CT80.1, HIWI, MIWI, PIWI
Chromosome Location
Chr.12: 130337887 - 130412708 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190971.1 NP_001177900.1 3 - 4 468 76
NM_004764.4 NP_004755.2 3 - 4 468 76
XM_011539002.2 XP_011537304.1 3 - 4 342 76
XM_011539003.2 XP_011537305.1 3 - 4 273 76
XM_011539004.2 XP_011537306.1 3 - 4 411 76
XM_011539005.1 XP_011537307.1 3 - 4 376 76
XM_017020229.1 XP_016875718.1 3 - 4 376 76
GenBank mRNA AB274731.1 - 2 - 3 197 76
AF104260.2 - 3 - 4 253 76
AF387507.1 - 3 - 4 260 76
AK093133.1 - 3 - 4 250 76
BC028581.2 - 3 - 4 265 76
DB072454.1 - 3 - 4 469 76

View Full Product Details