Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEK11
Entrez Gene ID
79858
Gene Name
NIMA related kinase 11
Gene Aliases
-
Chromosome Location
Chr.3: 131026850 - 131350465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146003.1 NP_001139475.1 6 - 7 907 75
NM_001321220.1 NP_001308149.1 6 - 7 907 75
NM_001321221.1 NP_001308150.1 8 - 9 1106 75
NM_001321223.1 NP_001308152.1 6 - 7 1025 75
NM_001321224.1 NP_001308153.1 5 - 6 741 75
NM_024800.4 NP_079076.3 7 - 8 980 75
NM_145910.3 NP_665917.1 7 - 8 980 75
XM_011513167.2 XP_011511469.1 7 - 8 1180 75
XM_011513173.1 XP_011511475.1 5 - 6 656 75
XM_011513174.1 XP_011511476.1 7 - 8 926 75
XM_011513175.1 XP_011511477.1 6 - 7 768 75
XM_011513181.1 XP_011511483.1 3 - 4 309 75
XM_017007205.1 XP_016862694.1 8 - 9 1283 75
XM_017007206.1 XP_016862695.1 8 - 9 1201 75
XM_017007207.1 XP_016862696.1 8 - 9 1417 75
XM_017007208.1 XP_016862697.1 7 - 8 1001 75
XM_017007209.1 XP_016862698.1 7 - 8 1210 75
XM_017007210.1 XP_016862699.1 7 - 8 1066 75
XM_017007211.1 XP_016862700.1 7 - 8 1001 75
XM_017007212.1 XP_016862701.1 7 - 8 979 75
XM_017007213.1 XP_016862702.1 7 - 8 979 75
XM_017007214.1 XP_016862703.1 6 - 7 1082 75
XM_017007215.1 XP_016862704.1 7 - 8 979 75
XM_017007216.1 XP_016862705.1 6 - 7 875 75
XM_017007218.1 XP_016862707.1 9 - 10 1184 75
XM_017007219.1 XP_016862708.1 6 - 7 678 75
XM_017007220.1 XP_016862709.1 7 - 8 1001 75
XM_017007221.1 XP_016862710.1 7 - 8 1001 75
XM_017007222.1 XP_016862711.1 7 - 8 1001 75
XM_017007224.1 XP_016862713.1 8 - 9 1045 75
XM_017007225.1 XP_016862714.1 8 - 9 1058 75
XM_017007226.1 XP_016862715.1 8 - 9 1322 75
XM_017007227.1 XP_016862716.1 6 - 7 802 75
XM_017007228.1 XP_016862717.1 4 - 5 417 75
XM_017007231.1 XP_016862720.1 6 - 7 780 75
XM_017007232.1 XP_016862721.1 7 - 8 985 75
XM_017007233.1 XP_016862722.1 6 - 7 735 75
GenBank mRNA AB071996.1 - 7 - 8 947 75
AB071997.1 - 7 - 8 947 75
AK027148.1 - 2 - 3 173 75
AK293261.1 - 5 - 6 656 75
AL833472.1 - 6 - 7 878 75
BC028587.2 - 6 - 7 1024 75

View Full Product Details