The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015000.3 | NP_055815.1 | 6 - 7 | 718 | 121 |
XM_005253342.3 | XP_005253399.1 | 5 - 6 | 597 | 121 | |
XM_006719058.3 | XP_006719121.1 | 7 - 8 | 699 | 121 | |
XM_006719059.3 | XP_006719122.1 | 7 - 8 | 678 | 121 | |
XM_011520613.2 | XP_011518915.1 | 5 - 6 | 412 | 121 | |
XM_017019036.1 | XP_016874525.1 | 6 - 7 | 674 | 121 | |
GenBank mRNA | AB023182.2 | - | 6 - 7 | 803 | 121 |
AK291055.1 | - | 6 - 7 | 608 | 121 | |
AK304755.1 | - | 4 - 5 | 302 | 121 | |
BC028603.1 | - | 6 - 7 | 692 | 121 |