The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024843.1 | NP_001020014.1 | 8 - 9 | 1025 | 56 |
NM_001162501.1 | NP_001155973.1 | 6 - 7 | 3175 | 56 | |
GenBank mRNA | AK294519.1 | - | 6 - 7 | 3166 | 56 |
AK304087.1 | - | 2 - 3 | 501 | 56 | |
BC028626.2 | - | 8 - 9 | 958 | 56 | |
BX647630.1 | - | 2 - 3 | 2218 | 56 |