Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD7
Entrez Gene ID
23424
Gene Name
tudor domain containing 7
Gene Aliases
CATC4, PCTAIRE2BP, TRAP
Chromosome Location
Chr.9: 97412020 - 97496125 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302884.1 NP_001289813.1 9 - 10 1953 107
NM_014290.2 NP_055105.2 10 - 11 2166 107
XM_006717026.3 XP_006717089.1 9 - 10 1456 107
GenBank mRNA AB025254.1 - 1 - 2 51 107
AK294488.1 - 9 - 10 1879 107
AK301954.1 - 3 - 4 534 107
AK310227.1 - 2 - 3 422 107
AK314853.1 - 10 - 11 2078 107
BC028694.1 - 10 - 11 2165 107

View Full Product Details