The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243533.1 | NP_001230462.1 | 2 - 3 | 327 | 75 |
XM_005245743.4 | XP_005245800.1 | 2 - 3 | 396 | 75 | |
GenBank mRNA | AF411102.1 | - | 2 - 3 | 267 | 75 |
BC028713.1 | - | 2 - 3 | 269 | 75 |