The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015141.3 | NP_055956.1 | 7 - 8 | 1158 | 80 |
XM_006713068.2 | XP_006713131.1 | 6 - 7 | 895 | 80 | |
GenBank mRNA | AK074221.1 | - | 4 - 5 | 557 | 80 |
AK125404.1 | - | 2 - 3 | 881 | 80 | |
AK292808.1 | - | 7 - 8 | 991 | 80 | |
BC006168.1 | - | 5 - 6 | 682 | 80 | |
BC028726.2 | - | 7 - 8 | 1017 | 80 | |
D42047.1 | - | 7 - 8 | 1138 | 80 |