The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130058.1 | NP_001123530.1 | 14 - 15 | 1228 | 106 |
NM_001320283.1 | NP_001307212.1 | 12 - 13 | 1189 | 106 | |
NM_001320285.1 | NP_001307214.1 | 16 - 17 | 1388 | 106 | |
NM_001320287.1 | NP_001307216.1 | 15 - 16 | 1321 | 106 | |
NM_152697.4 | NP_689910.2 | 14 - 15 | 1228 | 106 | |
XM_006710445.3 | XP_006710508.1 | 12 - 13 | 1115 | 106 | |
XM_011540986.2 | XP_011539288.1 | 13 - 14 | 1206 | 106 | |
XM_011540987.2 | XP_011539289.1 | 15 - 16 | 1283 | 106 | |
XM_017000607.1 | XP_016856096.1 | 13 - 14 | 1254 | 106 | |
XM_017000608.1 | XP_016856097.1 | 13 - 14 | 1253 | 106 | |
XM_017000609.1 | XP_016856098.1 | 16 - 17 | 2572 | 106 | |
XM_017000610.1 | XP_016856099.1 | 16 - 17 | 1650 | 106 | |
XM_017000611.1 | XP_016856100.1 | 14 - 15 | 1335 | 106 | |
GenBank mRNA | AK091400.1 | - | 2 - 3 | 168 | 106 |
AK093170.1 | - | 14 - 15 | 1230 | 106 | |
AK128063.1 | - | 15 - 16 | 1281 | 106 | |
AK296924.1 | - | 13 - 14 | 1267 | 106 | |
AK296941.1 | - | 12 - 13 | 1088 | 106 | |
AK299573.1 | - | 12 - 13 | 1189 | 106 | |
BC028743.2 | - | 14 - 15 | 1201 | 106 | |
BC034580.1 | - | 16 - 17 | 1367 | 106 | |
BC051740.1 | - | 16 - 17 | 1367 | 106 |