The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136052.2 | NP_001129524.1 | 15 - 16 | 2012 | 90 |
NM_001199752.2 | NP_001186681.1 | 15 - 16 | 2109 | 90 | |
NM_001199753.1 | NP_001186682.1 | 15 - 16 | 2045 | 90 | |
NM_152359.2 | NP_689572.1 | 14 - 15 | 1976 | 90 | |
NR_108072.1 | 16 - 17 | 2207 | 90 | ||
XM_005258505.2 | XP_005258562.1 | 15 - 16 | 2082 | 90 | |
XM_005258506.4 | XP_005258563.1 | 14 - 15 | 1998 | 90 | |
XM_006723009.3 | XP_006723072.1 | 12 - 13 | 1814 | 90 | |
XM_011526438.2 | XP_011524740.1 | 15 - 16 | 2138 | 90 | |
XM_011526439.2 | XP_011524741.1 | 15 - 16 | 2202 | 90 | |
XM_017026267.1 | XP_016881756.1 | 14 - 15 | 1872 | 90 | |
XM_017026268.1 | XP_016881757.1 | 13 - 14 | 1715 | 90 | |
XM_017026269.1 | XP_016881758.1 | 13 - 14 | 1825 | 90 | |
XM_017026270.1 | XP_016881759.1 | 13 - 14 | 1593 | 90 | |
XM_017026271.1 | XP_016881760.1 | 13 - 14 | 1682 | 90 | |
GenBank mRNA | AF357970.1 | - | 14 - 15 | 1934 | 90 |
AK074389.1 | - | 7 - 8 | 955 | 90 | |
AK096516.1 | - | 16 - 17 | 2203 | 90 | |
AK289713.1 | - | 15 - 16 | 2101 | 90 | |
AK290092.1 | - | 15 - 16 | 2017 | 90 | |
AL831876.1 | - | 5 - 6 | 607 | 90 | |
BC029104.1 | - | 15 - 16 | 1976 | 90 |