The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152338.3 | NP_689551.2 | 3 - 4 | 261 | 64 |
XM_011545921.2 | XP_011544223.1 | 3 - 4 | 340 | 64 | |
GenBank mRNA | AB092813.1 | - | 2 - 3 | 187 | 64 |
AK125559.1 | - | 3 - 4 | 219 | 64 | |
AK312002.1 | - | 2 - 3 | 209 | 64 | |
BC029149.1 | - | 2 - 3 | 193 | 64 | |
BC150656.1 | - | 3 - 4 | 232 | 64 | |
CK904265.1 | - | 3 - 4 | 261 | 64 |