The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152311.3 | NP_689524.1 | 2 - 3 | 575 | 96 |
GenBank mRNA | BC029478.1 | - | 2 - 3 | 575 | 96 |
BM830163.1 | - | 2 - 3 | 328 | 96 | |
BX640823.1 | - | 1 - 2 | 855 | 96 |