Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFSD8
Entrez Gene ID
256471
Gene Name
major facilitator superfamily domain containing 8
Gene Aliases
CCMD, CLN7
Chromosome Location
Chr.4: 127917805 - 127965984 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152778.2 NP_689991.1 11 - 12 1266 107
XM_005262893.1 XP_005262950.1 10 - 11 1180 107
XM_005262896.1 XP_005262953.1 9 - 10 1033 107
XM_005262897.2 XP_005262954.1 8 - 9 979 107
XM_011531830.1 XP_011530132.1 9 - 10 1066 107
XM_011531831.1 XP_011530133.1 7 - 8 865 107
GenBank mRNA AK074564.1 - 10 - 11 1115 107
AK294957.1 - 10 - 11 1108 107
AK315596.1 - 10 - 11 1112 107
BC029503.1 - 11 - 12 1231 107

View Full Product Details