The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270396.1 | NP_001257325.1 | 1 - 2 | 274 | 106 |
NM_152436.2 | NP_689649.1 | 1 - 2 | 274 | 106 | |
GenBank mRNA | AK093175.1 | - | 1 - 2 | 264 | 106 |
BC016749.1 | - | 1 - 2 | 281 | 106 | |
BC029557.1 | - | 1 - 2 | 235 | 106 |