Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAALC-AS2
Entrez Gene ID
157556
Gene Name
BAALC antisense RNA 2
Gene Aliases
BAALCOS, C8orf56
Chromosome Location
Chr.8: 103132963 - 103141342 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027071.1 1 - 2 89 68
GenBank mRNA BC029562.1 - 1 - 2 89 68

View Full Product Details