Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA3D
Entrez Gene ID
223117
Gene Name
semaphorin 3D
Gene Aliases
Sema-Z2, coll-2
Chromosome Location
Chr.7: 84995192 - 85187176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152754.2 NP_689967.2 10 - 11 1231 79
XM_011515960.1 XP_011514262.1 12 - 13 1541 79
XM_011515961.2 XP_011514263.1 10 - 11 1066 79
XM_017011873.1 XP_016867362.1 11 - 12 1730 79
GenBank mRNA AY358937.1 - 11 - 12 1297 79
BC029590.1 - 3 - 4 317 79

View Full Product Details