Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA3D
Entrez Gene ID
223117
Gene Name
semaphorin 3D
Gene Aliases
Sema-Z2, coll-2
Chromosome Location
Chr.7: 84995192 - 85187176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152754.2 NP_689967.2 12 - 13 1540 74
XM_011515960.1 XP_011514262.1 14 - 15 1850 74
XM_011515961.2 XP_011514263.1 12 - 13 1375 74
XM_017011873.1 XP_016867362.1 13 - 14 2039 74
GenBank mRNA AY358937.1 - 13 - 14 1606 74
BC029590.1 - 5 - 6 626 74

View Full Product Details