Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RICTOR
Entrez Gene ID
253260
Gene Name
RPTOR independent companion of MTOR complex 2
Gene Aliases
AVO3, PIA, hAVO3
Chromosome Location
Chr.5: 38937920 - 39074421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285439.1 NP_001272368.1 5 - 6 421 85
NM_001285440.1 NP_001272369.1 5 - 6 421 85
NM_152756.4 NP_689969.2 5 - 6 421 85
XM_006714463.3 XP_006714526.1 5 - 6 434 85
XM_011514005.2 XP_011512307.1 5 - 6 434 85
XM_011514006.2 XP_011512308.1 4 - 5 238 85
XM_017009311.1 XP_016864800.1 4 - 5 354 85
XM_017009312.1 XP_016864801.1 4 - 5 354 85
XM_017009313.1 XP_016864802.1 4 - 5 375 85
XM_017009314.1 XP_016864803.1 5 - 6 412 85
XM_017009315.1 XP_016864804.1 5 - 6 412 85
GenBank mRNA AK128358.1 - 4 - 5 354 85
AY515854.1 - 5 - 6 417 85
BC029608.1 - 5 - 6 412 85
BC137163.1 - 5 - 6 414 85
BC137164.1 - 5 - 6 414 85
BC144509.1 - 5 - 6 414 85
BP245264.1 - 5 - 6 421 85
CR749280.1 - 5 - 6 402 85

View Full Product Details