The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015627.2 | NP_056442.2 | 5 - 6 | 649 | 84 |
XM_006710559.3 | XP_006710622.1 | 5 - 6 | 646 | 84 | |
XM_006710560.3 | XP_006710623.1 | 5 - 6 | 646 | 84 | |
XM_006710561.3 | XP_006710624.1 | 5 - 6 | 646 | 84 | |
XM_011541209.2 | XP_011539511.1 | 5 - 6 | 646 | 84 | |
XM_011541210.2 | XP_011539512.1 | 5 - 6 | 646 | 84 | |
XM_011541211.2 | XP_011539513.1 | 5 - 6 | 646 | 84 | |
XM_011541212.2 | XP_011539514.1 | 5 - 6 | 646 | 84 | |
XM_017000994.1 | XP_016856483.1 | 5 - 6 | 1591 | 84 | |
GenBank mRNA | AK024397.1 | - | 4 - 5 | 1007 | 84 |
AK091219.1 | - | 5 - 6 | 654 | 84 | |
AL117654.1 | - | 5 - 6 | 556 | 84 | |
BC029770.1 | - | 5 - 6 | 559 | 84 | |
DA930479.1 | - | 2 - 3 | 121 | 84 |