The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163560.2 | NP_001157032.1 | 11 - 12 | 1231 | 107 |
NM_152764.2 | NP_689977.2 | 11 - 12 | 1154 | 107 | |
GenBank mRNA | BC029829.1 | - | 7 - 8 | 756 | 107 |