The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173602.2 | NP_775873.2 | 28 - 29 | 3558 | 73 |
XM_005269044.1 | XP_005269101.1 | 28 - 29 | 3561 | 73 | |
XM_006719520.3 | XP_006719583.1 | 29 - 30 | 3534 | 73 | |
XM_006719521.3 | XP_006719584.1 | 28 - 29 | 3371 | 73 | |
XM_011538594.2 | XP_011536896.1 | 29 - 30 | 3587 | 73 | |
XM_011538595.2 | XP_011536897.1 | 27 - 28 | 3419 | 73 | |
XM_011538596.2 | XP_011536898.1 | 28 - 29 | 3374 | 73 | |
XM_017019699.1 | XP_016875188.1 | 29 - 30 | 3569 | 73 | |
XM_017019700.1 | XP_016875189.1 | 28 - 29 | 3543 | 73 | |
XM_017019701.1 | XP_016875190.1 | 28 - 29 | 3540 | 73 | |
XM_017019702.1 | XP_016875191.1 | 26 - 27 | 3390 | 73 | |
XM_017019703.1 | XP_016875192.1 | 27 - 28 | 3337 | 73 | |
XM_017019704.1 | XP_016875193.1 | 27 - 28 | 3345 | 73 | |
XM_017019705.1 | XP_016875194.1 | 27 - 28 | 3327 | 73 | |
GenBank mRNA | AB040896.2 | - | 19 - 20 | 2172 | 73 |
AK097369.1 | - | 10 - 11 | 1114 | 73 | |
AK126758.1 | - | 17 - 18 | 1929 | 73 | |
BC007671.2 | - | 12 - 13 | 1482 | 73 | |
BC030156.2 | - | 12 - 13 | 1482 | 73 |