Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C3orf20
Entrez Gene ID
84077
Gene Name
chromosome 3 open reading frame 20
Gene Aliases
-
Chromosome Location
Chr.3: 14673532 - 14773036 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184957.1 NP_001171886.1 9 - 10 1381 70
NM_001184958.1 NP_001171887.1 9 - 10 1486 70
NM_032137.4 NP_115513.4 9 - 10 1883 70
XM_006713341.2 XP_006713404.1 9 - 10 1883 70
XM_006713342.2 XP_006713405.1 9 - 10 1736 70
XM_006713343.2 XP_006713406.1 9 - 10 1883 70
XM_011534153.2 XP_011532455.1 9 - 10 3513 70
XM_011534154.2 XP_011532456.1 9 - 10 2948 70
XM_011534157.2 XP_011532459.1 5 - 6 660 70
XM_011534158.2 XP_011532460.1 9 - 10 1883 70
XM_017007310.1 XP_016862799.1 9 - 10 1883 70
GenBank mRNA AK131481.1 - 9 - 10 1595 70
AL136781.1 - 9 - 10 1301 70
AL834386.1 - 9 - 10 1835 70
BC030599.2 - 9 - 10 1444 70
BC038406.1 - 9 - 10 1803 70

View Full Product Details