The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303279.1 | NP_001290208.1 | 7 - 8 | 1132 | 87 |
NM_001303280.1 | NP_001290209.1 | 7 - 8 | 1132 | 87 | |
NM_015194.2 | NP_056009.1 | 7 - 8 | 1132 | 87 | |
XM_017024685.1 | XP_016880174.1 | 7 - 8 | 1103 | 87 | |
GenBank mRNA | AB018270.2 | - | 7 - 8 | 843 | 87 |
AK026920.1 | - | 4 - 5 | 428 | 87 | |
AK127942.1 | - | 7 - 8 | 1103 | 87 | |
BC146763.1 | - | 7 - 8 | 843 | 87 | |
BX537615.1 | - | 6 - 7 | 565 | 87 |