Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR88
Entrez Gene ID
126248
Gene Name
WD repeat domain 88
Gene Aliases
PQWD
Chromosome Location
Chr.19: 33132092 - 33175799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173479.3 NP_775750.3 1 - 2 351 77
NR_135565.1 1 - 2 351 77
XM_011526449.2 XP_011524751.1 1 - 2 345 77
XM_011526450.2 XP_011524752.1 1 - 2 345 77
XM_017026282.1 XP_016881771.1 1 - 2 345 77
XM_017026283.1 XP_016881772.1 1 - 2 345 77
GenBank mRNA AK131444.1 - 1 - 2 710 77
BC031227.1 - 1 - 2 351 77

View Full Product Details