The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173479.3 | NP_775750.3 | 3 - 4 | 555 | 74 |
NR_135565.1 | 3 - 4 | 555 | 74 | ||
XM_011526449.2 | XP_011524751.1 | 3 - 4 | 549 | 74 | |
XM_011526450.2 | XP_011524752.1 | 3 - 4 | 549 | 74 | |
XM_017026282.1 | XP_016881771.1 | 3 - 4 | 549 | 74 | |
XM_017026283.1 | XP_016881772.1 | 3 - 4 | 549 | 74 | |
GenBank mRNA | AK131444.1 | - | 3 - 4 | 914 | 74 |
BC031227.1 | - | 3 - 4 | 555 | 74 |