Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM24B
Entrez Gene ID
196792
Gene Name
family with sequence similarity 24 member B
Gene Aliases
-
Chromosome Location
Chr.10: 122849094 - 122879641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152644.2 NP_689857.2 2 - 3 304 89
GenBank mRNA AV762334.2 - 2 - 3 244 89
BC031343.1 - 2 - 3 225 89

View Full Product Details