The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318812.1 | NP_001305741.1 | 3 - 4 | 603 | 79 |
NM_001318813.1 | NP_001305742.1 | 3 - 4 | 705 | 79 | |
NM_001318814.1 | NP_001305743.1 | 4 - 5 | 787 | 79 | |
NM_001318815.1 | NP_001305744.1 | 3 - 4 | 548 | 79 | |
NM_001318816.1 | NP_001305745.1 | 3 - 4 | 873 | 79 | |
NM_173810.3 | NP_776171.1 | 2 - 3 | 791 | 79 | |
GenBank mRNA | AF451992.1 | - | 2 - 3 | 732 | 79 |
AF451993.1 | - | 2 - 3 | 732 | 79 | |
BC032123.2 | - | 2 - 3 | 791 | 79 | |
CN430374.1 | - | 3 - 4 | 486 | 79 |