The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152318.2 | NP_689531.2 | 1 - 2 | 148 | 90 |
GenBank mRNA | BC032326.1 | - | - | 132 | 90 |
BG829346.1 | - | 1 - 2 | 149 | 90 |