Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL6
Entrez Gene ID
89857
Gene Name
kelch like family member 6
Gene Aliases
-
Chromosome Location
Chr.3: 183487531 - 183555712 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130446.2 NP_569713.2 2 - 3 509 82
XM_011513274.2 XP_011511576.1 2 - 3 510 82
GenBank mRNA AF441792.1 - 2 - 3 421 82
AK097125.1 - 2 - 3 463 82
AK314473.1 - 2 - 3 463 82
BC032348.1 - 2 - 3 487 82
BP284594.1 - 2 - 3 508 82
BX648370.1 - 2 - 3 464 82

View Full Product Details