The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152540.3 | NP_689753.2 | 4 - 5 | 1446 | 70 |
XM_011534375.2 | XP_011532677.1 | 4 - 5 | 1482 | 70 | |
XM_011534376.2 | XP_011532678.1 | 4 - 5 | 1482 | 70 | |
XM_017007786.1 | XP_016863275.1 | 4 - 5 | 1482 | 70 | |
XM_017007787.1 | XP_016863276.1 | 4 - 5 | 1482 | 70 | |
GenBank mRNA | AB074276.1 | - | 4 - 5 | 1419 | 70 |
AK096833.1 | - | 4 - 5 | 1432 | 70 | |
AY299407.1 | - | 4 - 5 | 1446 | 70 | |
BC012458.1 | - | 4 - 5 | 1212 | 70 | |
BC021137.1 | - | 4 - 5 | 1403 | 70 | |
BC032453.1 | - | 4 - 5 | 1444 | 70 |