The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281731.1 | NP_001268660.1 | 9 - 10 | 1309 | 116 |
NM_147190.3 | NP_671723.1 | 8 - 9 | 1108 | 116 | |
NR_104035.1 | 7 - 8 | 1050 | 116 | ||
XM_005269220.2 | XP_005269277.1 | 8 - 9 | 944 | 116 | |
XM_006719687.3 | XP_006719750.1 | 8 - 9 | 944 | 116 | |
XM_011538975.2 | XP_011537277.1 | 8 - 9 | 944 | 116 | |
XM_017020202.1 | XP_016875691.1 | 8 - 9 | 868 | 116 | |
XM_017020203.1 | XP_016875692.1 | 9 - 10 | 1232 | 116 | |
XM_017020205.1 | XP_016875694.1 | 8 - 9 | 797 | 116 | |
GenBank mRNA | AK026014.1 | - | 8 - 9 | 914 | 116 |
AK058033.1 | - | 6 - 7 | 1500 | 116 | |
AK298924.1 | - | 7 - 8 | 862 | 116 | |
AK300937.1 | - | 9 - 10 | 1084 | 116 | |
BC032565.1 | - | 8 - 9 | 925 | 116 | |
BC033558.1 | - | 7 - 8 | 837 | 116 |