The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175063.5 | NP_778233.4 | 3 - 4 | 363 | 89 |
NM_206538.3 | NP_996261.1 | 3 - 4 | 363 | 89 | |
XM_006723163.3 | XP_006723226.1 | 3 - 4 | 357 | 89 | |
XM_011526818.2 | XP_011525120.1 | 3 - 4 | 357 | 89 | |
GenBank mRNA | AY194293.2 | - | 7 - 8 | 1321 | 89 |
AY358710.1 | - | 3 - 4 | 323 | 89 | |
AY761095.1 | - | 3 - 4 | 341 | 89 | |
AY761097.1 | - | 3 - 4 | 341 | 89 | |
BC032948.2 | - | 3 - 4 | 304 | 89 | |
BC035001.2 | - | 3 - 4 | 302 | 89 | |
BC062607.1 | - | 3 - 4 | 505 | 89 | |
BX366174.2 | - | 3 - 4 | 361 | 89 | |
HY107516.1 | - | 3 - 4 | 364 | 89 |