Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM72
Entrez Gene ID
493829
Gene Name
tripartite motif containing 72
Gene Aliases
MG53
Chromosome Location
Chr.16: 31214021 - 31225656 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK090695.1 - 1 - 2 118 114
AK131485.1 - 2 - 3 618 114
BC033211.1 - 2 - 3 603 114
DA058421.1 - 1 - 2 118 114

View Full Product Details