Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC28A
Entrez Gene ID
25901
Gene Name
coiled-coil domain containing 28A
Gene Aliases
C6orf80, CCRL1AP
Chromosome Location
Chr.6: 138773517 - 138793319 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015439.2 NP_056254.1 4 - 5 903 89
GenBank mRNA AJ420568.1 - 4 - 5 654 89
AK300672.1 - 4 - 5 654 89
AL050197.1 - 4 - 5 653 89
AY167571.1 - 4 - 5 921 89
BC004464.1 - 4 - 5 638 89
BC013019.2 - 4 - 5 635 89
BC080566.1 - 4 - 5 637 89
BC108717.1 - 4 - 5 826 89

View Full Product Details