Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VGLL4
Entrez Gene ID
9686
Gene Name
vestigial like family member 4
Gene Aliases
VGL-4
Chromosome Location
Chr.3: 11556067 - 11720768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284390.1 NP_001271319.1 1 - 2 372 52
NM_014667.3 NP_055482.2 1 - 2 372 52
XM_017007542.1 XP_016863031.1 1 - 2 319 52
GenBank mRNA AK130542.1 - 1 - 2 167 52
BC001514.2 - 1 - 2 137 52
BC003038.1 - 1 - 2 335 52
D50911.3 - 1 - 2 120 52
DA321008.1 - 1 - 2 350 52
DA383197.1 - 1 - 2 167 52
HY020255.1 - 1 - 2 372 52

View Full Product Details