Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VGLL4
Entrez Gene ID
9686
Gene Name
vestigial like family member 4
Gene Aliases
VGL-4
Chromosome Location
Chr.3: 11556067 - 11720768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128219.2 NP_001121691.1 2 - 3 731 63
NM_001284390.1 NP_001271319.1 4 - 5 775 63
NM_001284391.1 NP_001271320.1 2 - 3 448 63
NM_014667.3 NP_055482.2 3 - 4 642 63
XM_011534267.2 XP_011532569.1 4 - 5 417 63
XM_011534268.1 XP_011532570.1 2 - 3 745 63
XM_011534269.1 XP_011532571.1 2 - 3 439 63
XM_017007541.1 XP_016863030.1 4 - 5 440 63
XM_017007542.1 XP_016863031.1 3 - 4 589 63
GenBank mRNA AK130542.1 - 3 - 4 437 63
BC001514.2 - 3 - 4 407 63
BC003038.1 - 4 - 5 738 63
D50911.3 - 3 - 4 390 63
DA301780.1 - 2 - 3 508 63
DA383197.1 - 3 - 4 437 63
DR159250.1 - 2 - 3 365 63

View Full Product Details