The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278460.1 | NP_001265389.1 | 2 - 3 | 306 | 80 |
NM_001278461.1 | NP_001265390.1 | 1 - 2 | 266 | 80 | |
NM_014900.4 | NP_055715.3 | 1 - 2 | 266 | 80 | |
GenBank mRNA | AB023194.1 | - | 1 - 2 | 260 | 80 |
AK002054.1 | - | 2 - 3 | 208 | 80 | |
AK225849.1 | - | 2 - 3 | 163 | 80 | |
BC006264.2 | - | 2 - 3 | 525 | 80 | |
BC071588.1 | - | 1 - 2 | 268 | 80 | |
CB989062.1 | - | - | 295 | 80 |