The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079675.2 | NP_001073143.1 | 10 - 11 | 1255 | 70 |
NM_001261437.1 | NP_001248366.1 | 9 - 10 | 1144 | 70 | |
NM_001261438.1 | NP_001248367.1 | 9 - 10 | 973 | 70 | |
NM_001261439.1 | NP_001248368.1 | 2 - 3 | 399 | 70 | |
NM_001986.2 | NP_001977.1 | 10 - 11 | 1084 | 70 | |
XM_011524514.1 | XP_011522816.1 | 7 - 8 | 630 | 70 | |
XM_017024349.1 | XP_016879838.1 | 10 - 11 | 1031 | 70 | |
GenBank mRNA | AK290429.1 | - | 10 - 11 | 1046 | 70 |
AK298872.1 | - | 8 - 9 | 890 | 70 | |
AK299019.1 | - | 9 - 10 | 935 | 70 | |
AK312934.1 | - | 10 - 11 | 1043 | 70 | |
AK315961.1 | - | 9 - 10 | 1035 | 70 | |
BC016623.1 | - | 10 - 11 | 1026 | 70 | |
BF304101.1 | - | 2 - 3 | 398 | 70 | |
D12765.1 | - | 8 - 9 | 890 | 70 | |
U18018.1 | - | 10 - 11 | 1159 | 70 |