The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021937.4 | NP_068756.2 | 3 - 4 | 676 | 99 |
XM_017006975.1 | XP_016862464.1 | 3 - 4 | 633 | 99 | |
XM_017006976.1 | XP_016862465.1 | 3 - 4 | 633 | 99 | |
XM_017006977.1 | XP_016862466.1 | 3 - 4 | 633 | 99 | |
XM_017006978.1 | XP_016862467.1 | 4 - 5 | 456 | 99 | |
XM_017006979.1 | XP_016862468.1 | 3 - 4 | 945 | 99 | |
XM_017006980.1 | XP_016862469.1 | 3 - 4 | 677 | 99 | |
GenBank mRNA | AF268872.1 | - | 3 - 4 | 415 | 99 |
BC007933.2 | - | 3 - 4 | 554 | 99 |