Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST6GAL2
Entrez Gene ID
84620
Gene Name
ST6 beta-galactoside alpha-2,6-sialyltransferase 2
Gene Aliases
SIAT2, ST6GalII
Chromosome Location
Chr.2: 106801600 - 106887410 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142351.1 NP_001135823.1 3 - 4 1158 131
NM_001142352.1 NP_001135824.1 3 - 4 1228 131
NM_001322362.1 NP_001309291.1 3 - 4 1207 131
NM_032528.2 NP_115917.1 3 - 4 1228 131
NR_136313.1 2 - 3 243 131
XM_006712802.1 XP_006712865.1 3 - 4 1243 131
XM_011511999.2 XP_011510301.1 3 - 4 1340 131
XM_011512000.2 XP_011510302.1 3 - 4 1425 131
XM_011512001.2 XP_011510303.1 3 - 4 1425 131
XM_017005110.1 XP_016860599.1 3 - 4 1179 131
GenBank mRNA AB058780.1 - 3 - 4 1158 131
AB059555.1 - 3 - 4 1214 131
AJ512141.1 - 1 - 2 99 131
AK307784.1 - 3 - 4 1228 131
BC008680.1 - 3 - 4 1214 131

View Full Product Details