The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080658.1 | NP_542389.1 | 5 - 6 | 691 | 148 |
XM_017018549.1 | XP_016874038.1 | 6 - 7 | 882 | 148 | |
XM_017018550.1 | XP_016874039.1 | 6 - 7 | 916 | 148 | |
XM_017018551.1 | XP_016874040.1 | 6 - 7 | 837 | 148 | |
GenBank mRNA | AF359506.1 | - | 4 - 5 | 610 | 148 |
AY032593.1 | - | - | 671 | 148 | |
AY040761.1 | - | 5 - 6 | 671 | 148 | |
AY169233.1 | - | 5 - 6 | 691 | 148 | |
BC008689.1 | - | 5 - 6 | 691 | 148 |