The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146726.1 | NP_001140198.1 | 6 - 7 | 989 | 62 |
NM_138379.2 | NP_612388.2 | 7 - 8 | 1073 | 62 | |
XM_017010021.1 | XP_016865510.1 | 5 - 6 | 879 | 62 | |
GenBank mRNA | AA843555.1 | - | 2 - 3 | 62 | |
AK131025.1 | - | 3 - 4 | 331 | 62 | |
AK301337.1 | - | 6 - 7 | 960 | 62 | |
AK312746.1 | - | 7 - 8 | 1043 | 62 | |
BC008988.1 | - | 7 - 8 | 1073 | 62 | |
JX049980.1 | - | 7 - 8 | 1016 | 62 | |
KU178832.1 | - | 7 - 8 | 1016 | 62 | |
KU178833.1 | - | 6 - 7 | 935 | 62 |