The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243254.1 | NP_001230183.1 | 8 - 9 | 2168 | 106 |
NM_015213.3 | NP_056028.2 | 8 - 9 | 2168 | 106 | |
XM_005252832.1 | XP_005252889.1 | 8 - 9 | 2177 | 106 | |
GenBank mRNA | AB029014.1 | - | 8 - 9 | 2128 | 106 |
AK025499.1 | - | 3 - 4 | 779 | 106 | |
AK026007.1 | - | 1 - 2 | 244 | 106 | |
AK294016.1 | - | 7 - 8 | 1897 | 106 | |
AK295881.1 | - | 8 - 9 | 1929 | 106 | |
AK302478.1 | - | 5 - 6 | 1629 | 106 | |
AL117448.1 | - | 8 - 9 | 1924 | 106 | |
BC009354.2 | - | 4 - 5 | 896 | 106 | |
BC071596.1 | - | 9 - 10 | 1973 | 106 |