The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243254.1 | NP_001230183.1 | 20 - 21 | 3645 | 63 |
NM_015213.3 | NP_056028.2 | 20 - 21 | 3645 | 63 | |
XM_005252832.1 | XP_005252889.1 | 20 - 21 | 3654 | 63 | |
XM_011519953.2 | XP_011518255.1 | 15 - 16 | 1908 | 63 | |
GenBank mRNA | AB029014.1 | - | 20 - 21 | 3605 | 63 |
AK025499.1 | - | 15 - 16 | 2256 | 63 | |
AK125444.1 | - | 12 - 13 | 2950 | 63 | |
AK295881.1 | - | 20 - 21 | 3406 | 63 | |
AK302478.1 | - | 17 - 18 | 3106 | 63 | |
AL117448.1 | - | 20 - 21 | 3401 | 63 | |
BC009354.2 | - | 16 - 17 | 2373 | 63 | |
BC071596.1 | - | 21 - 22 | 3450 | 63 |