Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAUS8
Entrez Gene ID
93323
Gene Name
HAUS augmin like complex subunit 8
Gene Aliases
DGT4, HICE1, NY-SAR-48
Chromosome Location
Chr.19: 17049761 - 17075533 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011699.1 NP_001011699.1 7 - 8 593 68
NM_033417.1 NP_219485.1 7 - 8 596 68
XM_005260154.4 XP_005260211.1 6 - 7 429 68
GenBank mRNA AK295994.1 - 7 - 8 668 68
AY211919.1 - 7 - 8 498 68
BC004398.1 - 7 - 8 483 68
BC010176.2 - 1 - 2 32 68
BI829542.1 - 7 - 8 596 68
BM563712.1 - 7 - 8 477 68

View Full Product Details