The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080115.1 | NP_001073584.1 | 6 - 7 | 919 | 65 |
NM_001080116.1 | NP_001073585.1 | 7 - 8 | 883 | 65 | |
NM_001171611.1 | NP_001165082.1 | 7 - 8 | 1123 | 65 | |
XM_005269468.4 | XP_005269525.1 | 7 - 8 | 1084 | 65 | |
XM_011539195.2 | XP_011537497.1 | 8 - 9 | 1289 | 65 | |
XM_017015608.1 | XP_016871097.1 | 7 - 8 | 2983 | 65 | |
XM_017015609.1 | XP_016871098.1 | 8 - 9 | 842 | 65 | |
GenBank mRNA | AF276807.1 | - | 6 - 7 | 896 | 65 |
AF276808.1 | - | 8 - 9 | 1123 | 65 | |
AF276809.1 | - | 7 - 8 | 1100 | 65 | |
AK297871.1 | - | 7 - 8 | 875 | 65 | |
AK300692.1 | - | 8 - 9 | 791 | 65 | |
BC010929.2 | - | 7 - 8 | 780 | 65 | |
KU178543.1 | - | 7 - 8 | 755 | 65 | |
KU178544.1 | - | 6 - 7 | 896 | 65 | |
KU178545.1 | - | 5 - 6 | 528 | 65 |