The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032439.3 | NP_115815.2 | 1 - 2 | 372 | 63 |
GenBank mRNA | AK054956.1 | - | 1 - 2 | 176 | 63 |
AL834339.1 | - | 1 - 2 | 373 | 63 | |
BC011268.2 | - | 1 - 2 | 211 | 63 |