Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARS2
Entrez Gene ID
25973
Gene Name
prolyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
MT-PRORS, proRS
Chromosome Location
Chr.1: 54756898 - 54764553 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152268.3 NP_689481.2 1 - 2 88 64
GenBank mRNA AK289679.1 - 1 - 2 40 64
BC011758.2 - 1 - 2 49 64
DB004072.1 - 1 - 2 88 64

View Full Product Details